Would you like to log in again?
TWEEZERS STRIPPER 42 AWG CB STEEL 4.5IN
Supplier: EXCELTA CORPORATION
Tweezer 4.5in^ Tweezer, Stripper, Quality Level: 5stars, Shape: 70 deg Angulated, Material: Carbon Steel, Cutting Capacity: 42 AWG, Over All Length: 4.5in, Body Width: 0.393in, Body Height: 0.10in stripping tweezer will strip PTFE and other coated wire exactly to AWG gauge without nicking.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TWEEZERS STRIPPER 42 AWG CB STEEL 4.5IN 75880-812 15A-ST-PE-42 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty