Would you like to log in again?
CAP CRIMP BASIK WHT SEPTA P-SL RBR 11MM
Supplier: MicroSolv Technology Corp
Basik* Cap white septa 11mm^ Basik* Cap, crimp, silver aluminum with preslit, fitted white Silicone rubber/ red PTFE Soft-Guard septa. Use with snap or crimp top 12x32mm autosampler vials. Size: 11mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP CRIMP BASIK WHT SEPTA P-SL RBR 11MM 75880-958 9502C-E-43T-S Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty