Would you like to log in again?
Spatulas, Miniature, Aluminum Handle, Excelta Corp®
Supplier: Excelta
260C-ESD 260K-ESD 260A-ESD 260D-ESD 260AA-ESD 260E-ESD 260C-ESD 260B-ESD
76471-546EA 10.06 USD InStock
76471-546 89411-718 89411-726 75880-968 89411-722 89411-724 76243-464 89411-720
Spatulas, Miniature, Aluminum Handle, Excelta Corp®
Spatulas
Placing epoxy/lubricants in very small amounts.
  • Miniature design for accessibility under scope
  • Color coded width sizes for identification
  • Different tip widths to fit different applications
These mini-spatulas (oilers) all feature 2.5" long with aluminum handles. Mini-spatulas are ideal for placing epoxies or lubricants in very small places. They are all conductive. They are electrically conductive. The color coated handles identify the tip width.
Spatula shape: Flat end blade/straight handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty