Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices
Supplier: Molecular Devices
ID3-INJ-UPG ID3-STD ID3-STD ID3-INJ-UPG
75886-130EA 11107.84 USD InStock
75886-130 CA75886-130 CA75886-128 75886-128
SpectraMax® iD3 Multi-Mode Microplate Reader, Molecular Devices
Microplate Readers
Unparalleled performance on a personalized platform. The SpectraMax® iD3 Multi-Mode Microplate Reader is the cornerstone of a complete laboratory solution designed to expand the boundaries of research capabilities. With optimized reagents and the industry-leading data acquisition and analysis tool, SoftMax Pro® 7 Software, the SpectraMax® iD3 allows users to customize workflow to perfectly match their needs.
  • Use the touchscreen to easily set up methods, run experiments, or view tutorial videos
  • Personalize workflows with one-tap near-field communication (NFC) functionality
  • Push data to workstations, eliminating the need to retrieve data directly from the instrument
  • Validate the instrument and software with an extensive suite of tools
The SpectraMax® iD3 Multi-Mode Microplate Reader features a large, high-resolution touchscreen with an embedded software package that enables users to customize protocols, take advantage of preconfigured protocols, and run experiments without the need for a dedicated computer workstation
View data quickly using the large touchscreen, export to a USB drive, or analyze data using the industry-leading data acquisition and analysis tool, SoftMax Pro® 7 Software. QuickSync technology allows users to walk away from the SpectraMax® iD3 to focus on additional research. Data is automatically delivered to a workstation, eliminating the need to physically retrieve data from the instrument. Featuring orbital shaking, a four-monochromator optical pathway with high efficiency gratings, well scanning up to a 20x20 read matrix, spectral scanning and detection of plate formats from 6- to 384-wells, the SpectraMax® iD3 reader is a complete research solution.
Expand any lab’s capabilities to include flash applications such as dual luciferase and ATP assays with the SpectraMax® iD3 injector system with SmartInject™ Technology ensuring equal mixing across the plate for high-precision experiments. Engineered for high performance, the dual injector system can be used with any read mode and includes safeguard features such as overflow protection to save precious reagents and maximize performance for those critical experiments in the lab. SoftMax Pro® GxP Compliance Software extends Molecular Devices leading data acquisition and analysis solution into regulated laboratories working under GMP, GLP, 21 CFR Part 11, and other similar guidelines for secure electronic records.
A built-in near-field communication (NFC) reader offers access to custom protocols with a single tap, saving precious time. Pair NFC Tags with specific user profiles to gain access to personalized protocols and experiment results, eliminating the need to scroll through dozens of files. The superior optical system includes a xenon flash lamp and features an ultra-cooled photomultiplier tube (PMT) to –5 °C that reduces background noise for excellent sensitivity and a wide dynamic range. Simple-to-use temperature control provides adjustment to experimental conditions, from ambient up to 65 °C, expanding laboratory capabilities to include temperature sensitive assays.
Certifications: Meets ASTM standards. EN-342.
Ordering information: SpectraMax® iD3 Multi-Mode Microplate Reader comes standard with Absorbance, Fluorescence, and Luminescence as well as a superior optical system that includes a xenon flash lamp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty