Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DPP4 Mouse Monoclonal Antibody (CF660R) [clone: DPP4/910]

Supplier: Biotium

BNC610910-500 BNC610910-100
75891-552EA 547.44 USD InStock
75891-552 75891-550
Anti-DPP4 Mouse Monoclonal Antibody (CF660R) [clone: DPP4/910]
Antibodies
This antibody recognizes a glycoprotein of 110 kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®660R is a far-red fluorescent dye (Ex/Em 663/682 nm) with superior brightness and unrivaled photostability among spectrally similar dyes.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: Clone DPP4/910
Conjugation: CF660R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty