Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B3GAT1 Mouse Monoclonal Antibody [clone: HNK-1]

Supplier: Biotium

BNUM0136-50
75912-516EA 407.5 USD InStock
75912-516
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: HNK-1]
Antibodies
Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.

Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: Clone HNK-1
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty