Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: 158-4D3]

Supplier: Biotium

BNC470028-500 BNC470028-100
75912-994EA 197.58 USD InStock
75912-994 75912-996
Anti-PTPRC Mouse Monoclonal Antibody (CF647) [clone: 158-4D3]
Antibodies
This antibody recognizes a protein of 205 kDa-220 kDa, identified as CD45RA [Workshop V; Code CD45.38]. CD45RA is isoforms of the human leukocyte common antigen (CD45). Human CD45 contains three exons which encode peptide segments designated A, B and C, respectively. The differential splicing of the exons generates at least five isoforms, ABC, AB, BC, B and O. This antibody reacts with ABC and BC isoforms. CD45RA is expressed on 40-50% of peripheral CD4 T-cells, 50% of peripheral CD8 T-cells, B-cells, and leukemic B-cell lines. T-cells expressing CD45RA are naive or virgin T-cells. T-cells expressing CD45RO are memory T-cells. CD45RA and CD45RO define complementary, predominantly non-overlapping populations of resting peripheral T-cells. This MAb is useful in study on the subpopulation of CD4 or CD8 T-cells. It can especially be used to differentiate T-cell lymphomas (CD45RO ve) from B cell lymphomas (CD45RA ve).

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: Clone 158-4D3
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty