Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD46 Mouse Monoclonal Antibody (Biotin) [clone: 197-2B1]

Supplier: Biotium

BNCB0931-500 BNCB0931-100
75913-664EA 547.44 USD InStock
75913-664 75913-662
Anti-CD46 Mouse Monoclonal Antibody (Biotin) [clone: 197-2B1]
Antibodies
This antibody recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor for pathogenic Neisseria. This MAb can be applied to test complement activation in pseudo-allergic reactions to acetylsalicylic acid and to test for measles virus infection of cells.

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: Clone 197-2B1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty