Would you like to log in again?
Westex DH Flame Resistant Coverall, Navy Blue, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C065DH13-40-30 C065DH13-64-30 C065DH13-60-30 C065DH13-56-30 C065DH13-48-30 C065DH13-36-30 C065DH13-44-30 C065DH13-52-30
75923-438EA 211.24 USD InStock
75923-438 75923-440 75927-796 75923-444 75927-800 75927-798 75923-442 75923-436
Westex DH Flame Resistant Coverall, Navy Blue, Lakeland Industries
Workwear
Inherently flame resistant coveralls for primary protection from flash fire and electrical arc flash.
  • Inherently flame resistant fabric will not lose flame resistant properties
  • Pleated back and elastic waist for fit and comfort
  • Flared leg cuffs for ease of donning and doffing
Westex DH Flame Resistant Coveralls are dual certified for both flash fire and electrical arc flash protection. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier and sleeve cuffs feature adjustable snap closures.
Certifications: NFPA 2112, UL certified for flash fire protection. NFPA 70E, CAT2 electrical arc flash protection.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty