Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7 oz. 100% Flame Resistant Cotton Coverall, Navy, Lakeland Industries

Supplier: Lakeland Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

NC070FRC13-XL30 NC070FRC13-LG30 NC070FRC13-4X30 NC070FRC13-5X30 NC070FRC13-2X30 NC070FRC13-SM30 NC070FRC13-MD30 NC070FRC13-3X30
75923-658EA 111.79 USD InStock
75923-658 75923-660 75923-664 75923-520 75923-662 75923-524 75923-666 75923-522
7 oz. 100% Flame Resistant Cotton Coverall, Navy, Lakeland Industries
Workwear
Economical Flame Resistant Coverall for primary protection from flash fire and electrical arc flash.

  • Lightweight 7 oz. 100% Flame Resistant Cotton fabric
  • NFPA 2112, UL Certified for Flash Fire Protection
  • NFPA 70E, CAT2 Electrical Arc Flash Protection
  • Pleated back and elastic waist for fit and comfort
  • Flared leg cuffs for ease of donning and doffing

Lakeland Economical and Lightweight 7 oz. 100% Flame Resistant Cotton Coverall, dual certified for both flash fire and electrical arc flash protection. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier, and sleeve cuffs feature adjustable snap closures.

Certifications: NFPA 2112. NFPA 70E, CAT2.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty