Would you like to log in again?
7 oz. 88/12 Flame Resistant Coverall, Khaki, Lakeland Industries
Supplier: Lakeland Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C07120-3X30 C07120-5X30 C07120-MD30 C07120-XL30 C07120-SM30 C07120-LG30 C07120-2X30 C07120-4X30
75923-648EA 133.86 USD InStock
75923-648 75923-652 75923-650 75923-646 75923-502 75923-656 75923-644 75923-654
7 oz. 88/12 Flame Resistant Coverall, Khaki, Lakeland Industries
Workwear
Flame Resistant Coverall for primary protection from flash fire and electrical arc flash.
  • Lightweight 7 oz. 88/12 Flame Resistant fabric
  • NFPA 2112, UL Certified for Flash Fire Protection
  • NFPA 70E, CAT2 Electrical Arc Flash Protection
  • Pleated back and elastic waist for fit and comfort
  • Flared leg cuffs for ease of donning and doffing
Lakeland 7 oz. 88/12 Flame Resistant Coverall, dual certified for both flash fire and electrical arc flash protection. Designed for fit and comfort with pleated back, elastic waist, and plenty of pockets. Flared leg openings make donning and doffing easier, and sleeve cuffs feature adjustable snap closures.
Certifications: NFPA 2112. NFPA 70E, CAT2.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty