Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TransforMax™EPI300™ Electrocompetent and Chemically Competent E. coli, Biosearch Technologies
Supplier: Biosearch Technologies
TransforMax™
EC300150 CCIS125 C300C105 EC300110
75927-928EA 786.43 USD InStock
75927-928 75927-886 75927-874 75927-930
TransforMax™EPI300™ Electrocompetent and Chemically Competent E. coli, Biosearch Technologies
Competent Cells
Clone large inserts and obtain high colony counts from Gibson Assembly® reactions.
  • Clone large inserts and transform large plasmids - up to at least 145 kb plasmid DNA
  • Increase colony count after transformation of Gibson Assembly(R) reactions with high efficiency electrocompetent cells - endorsed by SGI-DNA
  • Generate clones with inducible copy number using CopyControl™ vectors
  • Choose from chemically competent format (>5 x 108 cfu/µg pUC19 DNA) or electrocompetent format (>1 x 1010 cfu/µg pUC19 DNA)
Delivery information: TransforMax™ EPI300™ Chemically Competent E. coli kit contains ten tubes, each containing 50 µL of cells (enough cells for 10 transformations), CopyControl™ Induction Solution and pUC19 control DNA.
TransforMax™ EPI300™ Electrocompetent E. coli kit contains ten or fifty tubes, each containing 100 µL of cells (enough cells for 20 or 100 transformations), CopyControl™ Induction Solution and pUC19 control DNA.
The CopyControl™ Induction Solution is provided at a 1000X concentration and is filter sterilized.
Caution: For research use only. Not for human or diagnostic use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty