Would you like to log in again?
TransforMax™EC100™ Electrocompetent and Chemically Competent E. coli, Biosearch Technologies
Supplier: Biosearch Technologies
TransforMax™
CC02810 EC10010
75927-880EA 301.84 USD InStock
75927-880 75927-926
TransforMax™EC100™ Electrocompetent and Chemically Competent E. coli, Biosearch Technologies
Competent Cells
Clone large plasmids with high efficiency.
  • Clone large inserts and transform large plasmids - up to at least 145 kb plasmid DNA
  • Select convenience with chemically competent format: >5 x 108 cfu/µg pUC19 DNA
  • Choose electrocompetent format for higher efficiencies: >1 x 1010 cfu/µg pUC19 DNA
  • Ideal strain for general cloning, subcloning and plasmid preparation due to endA and recA mutation
The highly versatile TransforMax™ EC100™ <italic>E
EC100 competent cells are recombination minus (recA) and endonuclease minus (endA), for high yield, high quality plasmid DNA preparations. They are also blue/white screening capable (lacZΔM15) and restriction minus [mcrA, Δ(mrr-hsdRMS-mcrBC)] for cloning methylated DNA.
Genotype: F- mcrA Δ(mrr-hsdRMS-mcrBC) Φ80dlacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara, leu)7697 galU galK λ- rpsL (StrR) nupG.
coli</italic> competent cells are ideal for most cloning applications. The cells provide very high transformation efficiency when tested against a wide range of supercoiled DNAs as well as DNA directly from a ligation reaction, including BAC clones.
Delivery information: TransforMax™ EC100™ Chemically Competent E. coli kit contains ten tubes, each containing 50 µL of cells (enough cells for 10 transformations) and pUC19 control DNA.
TransforMax™ EC100™ Electrocompetent E. coli kit contains ten tubes, each containing 100 µL of cells (enough cells for 20 transformations) and pUC19 control DNA.
Caution: For research use only. Not for human or diagnostic use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty