Would you like to log in again?
Ribonucleoside-5’-Triphosphate Solutions & 2’-Fluorine-Nucleoside-5’-Triphosphate Solutions, Biosearch Technologies
Supplier: Biosearch Technologies
RA02825 R2F110U RN02825 R2F110C
75927-966EA 546.17 USD InStock
75927-966 75935-550 75927-962 75935-552
Ribonucleoside-5’-Triphosphate Solutions & 2’-Fluorine-Nucleoside-5’-Triphosphate Solutions, Biosearch Technologies
Microarray Reagents
Ready-to-use NTP and dNTP solutions for all your RNA synthesis needs.
  • Ideal for use in RNA synthesis reactions
  • Provided as RNase- and DNase-free
The NTP Solutions are ideal for use in RNA synthesis reactions
2’-Fluorine-dCTP and 2’-Fluorine-dUTP are efficiently incorporated into RNA transcripts by in vitro transcription reactions with Durascribe® T7 RNA Polymerase. Complete substitution of the 2′-Fluorine-dCTP and 2′-Fluorine-dUTP for the canonical CTP and UTP in a DuraScribe® in vitro transcription reaction yields “DuraScript RNA” that is completely resistant to digestion by RNase A. DuraScript RNA is suitable for RNAi and RNA aptamer applications. These nucleotides are supplied as neutralized solutions at 50 mM each (1 µmol/tube).
The NTP and 2’-Fluorine-Nucleoside-5’-Triphosphate solutions are provided as RNase- and DNase-free, neutral solutions that meet strict spectrophotometric criteria and are tested in functional assays.
The ATP Solution is available separately and is suitable for use as a cofactor with T4 DNA Ligase for ligating DNA molecules and T4 Polynucleotide Kinase for phosphorylating the 5′ ends of DNA or RNA molecules, and for RNA tailing applications. The ATP Solution is provided as a 100 mM solution (25 µmol/tube).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty