Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GP6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BN2
75928-186EA 518 USD InStock
75928-186
Anti-GP6 Rabbit Polyclonal Antibody
Antibodies
Glycoprotein VI (GP6) is a 58kD platelet membrane glycoprotein that plays a crucial role in the collagen-induced activation and aggregation of platelets. It is uniquely expressed by cells of the megakaryocytic/platelet lineage, and is a member of the immunoglobulin gene superfamily, closely related to Fc receptor gamma chain (FcRgamma) and natural killer receptors. Glycoprotein VI plays a key role in platelet procoagulant activity and subsequent thrombin and fibrin formation. This procoagulant function may contribute to arterial and venous thrombus formation. The signaling pathway involves the FcRgamma, the Src kinases (likely Fyn/Lyn), the adapter protein LAT and leads to the activation of phospholipase C gamma2. GPVI deficiency can result in bleeding disorders. Further study should reveal the extent of GPVI involvement in thrombotic disease and allow the development of alternative anti-thrombotic compounds.

Anti-GPVI Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 37 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: GP6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty