Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIK4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BN8
75928-214EA 518 USD InStock
75928-214
Anti-GRIK4 Rabbit Polyclonal Antibody
Antibodies
Grik4 codes for the KA1 subunit of kainate-type ionotropic glutamate receptors which are critical regulators of network activity that act by modifying neuronal excitability, directly and indirectly, through GABAergic interneurons. Five subunits can assemble to form kainate receptors (KARs): GluR5 (coded by Grik1), GluR6 (coded by Grik2), and GluR7 (coded by Grik3) are the low-affinity subunits, and KA1 and KA2 are the high-affinity subunits. In the adult brain, KARs are located pre- and postsynaptically on pyramidal cells and on interneurons. Kainate receptors on GABA-containing interneurons enhance GABA release and thereby downregulate glutamatergic signaling. KARs have been implicated in numerous psychiatric disorders. Case control studies show significant association of Grik4 with both schizophrenia and bipolar disorder.

Anti-Grik4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 107 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: GRIK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty