Would you like to log in again?
Anti-GST Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Rockland Immunochemicals
200-102-C45S 200-102-C45
75928-276EA 487 USD InStock
75928-276 75928-278
Anti-GST Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Anti-GST Antibody Fluorescein Conjugated
  • Purified IgG Fraction
  • FITC
  • multiplex, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting
Rockland produces a wide range of GST antibodies in our laboratories. Select GST antibodies from several monoclonal and/or polyclonal GST antibodies listed below. Select appropriate GST antibodies for your research by isotype, epitope, applications and species reactivity. GST (Glutathione-S-Transferase) is a protein expression tag commonly used in molecular biology. Anti-GST will react with synthetic construct present in most known GST containing cloning or expression vectors. GST is responsible for the conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. The amino acid sequence GST is highly conserved in most organisms including mammals. GST exists as a 26 kDa homodimer.
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty