Would you like to log in again?
Guinea Pig Blood In Heparin Sodium, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R505-0100 R505-0050
75928-284EA 898 USD InStock
75928-284 75928-286
Guinea Pig Blood In Heparin Sodium, Rockland Immunochemicals
Blood
Guinea Pig Blood In Heparin Sodium
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Guinea pig whole blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Guinea pig red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty