Would you like to log in again?
Guinea Pig Buffy Coat Preparation, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R605-0100
75928-292EA 1186 USD InStock
75928-292
Guinea Pig Buffy Coat Preparation, Rockland Immunochemicals
Blood
Guinea Pig Buffy Coat Preparation
  • aseptically collected and prepared to order
  • enriched for leukocytes (WBC)
Blood is a complex mixture of cells, proteins, metabolites and many other substances.  About 56% of human blood volume is comprised of cells, more than 99% of which are erythrocytes.  Human erythrocytes and thrombocytes (platelets, 0.5% blood components) do not contain nuclei.  Leukocytes (0.3% of blood components), also referred to as peripheral blood mononuclear leukocytes or PBMC, do contain nuclei.  Blood samples may vary widely in leukocyte (granulocytes, lymphocytes and monocytes) concentration depending on the health of the donor. Healthy blood contains less than 10E7 leukocytes per ml, while blood from an infected donor may have ten fold higher leukocyte concentrations.
Buffy coat preparations are suitable for use as a source of nucleated PBMC. Buffy coat preparations are separated after a single low-speed centrifugation to remove the platelet rich plasma, buffy coat layer and erythrocytes (RBC). Buffy coat preparations are generally enriched for leukocytes (WBC) and contain trace amounts of red cells and other blood components. If desired, you may further purify this preparation of leukocytes by sedimentation using Percoll™ gradient centrifugation. This product is prepared in a buffer composed of 20.5 mg/ml Dextrose, 4.2mg/ml Sodium Chloride, 8.0 mg/ml Citric Acid Tri-sodium, adjusted to pH 6.1.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty