Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X11
75928-438EA 518 USD InStock
75928-438
Anti-HDAC2 Rabbit Polyclonal Antibody
Antibodies
Histone Deacetylase 2 (HDAC2) is part of a family of histone deacetylases that are responsible for deacetylation of lysine residues in the histone core. HDAC2 is classified as a class I histone deacetylase and is ubiquitously expressed throughout the body. It has been shown that HDAC2 plays an important role in cardiac hypertrophy. Phosphorylation of ser394 is responsible for the hypertrophy-associated activation of HDAC2, whereas intrinsic basal activity is maintained by phosphorylation of ser422 and ser424. HDAC2 pS394 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser394 HDAC2 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~55 kDa HDAC2 protein phosphorylated at Ser394 in Western blots. Immunolabeling is completely blocked by lambda-Phosphatase treatment (30 minutes, 800units/1mg protein). Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HDAC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty