Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IW52_sMgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BQ8
75928-446EA 518 USD InStock
75928-446
Anti-IW52_sMgp1 Rabbit Polyclonal Antibody
Antibodies
The Heartland virus is a newly identified member of the phlebovirus genus of the Bunyaviridae family. Initial reports indicate that symptoms may include fever, fatigue, diarrhea, thrombocytopenia, and leukopenia (1). The Bunyaviridae family of viruses are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity (2). The glycoprotein RNA encodes a polyprotein that is cleaved into the G1 and G2 proteins.

Anti-Heartland Virus Glycoprotein 2 Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 117 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IW52_sMgp1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty