Would you like to log in again?
HeLa Cell Nuclear Extract Etoposide Stimulated, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
W09-001-A83
75928-452EA 210 USD InStock
75928-452
HeLa Cell Nuclear Extract Etoposide Stimulated, Rockland Immunochemicals
Cells
HeLa Cell Nuclear Extract Etoposide Stimulated
  • Ready-to-use lysates
  • positive controls
  • treated lysated
  • nuclear lysates
Ready-to-use HeLa Cell Nuclear Extract Etoposide Stimulated produced by Rockland Immunochemicals are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality, protein integrity and lot-to-lot reproducibility. All extracts are tested by SDS-PAGE using 4-20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Ready-to-use nuclear extracts are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Nuclear extracts are supplied in denaturing buffer without dissociating agents. Heat nuclear extract to 95° C for 5 minutes and rapidly cool. If dissociating conditions are desired add reducing agent prior to heating. The recommended loading volume per lane is 10-30 ul depending on the size format of your gel.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty