Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-BR9
75928-476EA 518 USD InStock
75928-476
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Histones are the main constituents of the protein part of chromosomes of eukaryotic cells. They are rich in the amino acids arginine and lysine and have been greatly conserved during evolution. Histones pack the DNA into tight masses of chromatin. Two core histones of each class H2A, H2B, H3 and H4 assemble and are wrapped by 146 base pairs of DNA to form one octameric nucleosome. Histones play a internal role in the regulation of transcription, DNA repair, DNA replication and chromosomal stability. These different functions are established via a complex set of post-translational modifications which either directly or indirectly alter chromatin structure and DNA accessibility to facilitate transcriptional activation or repression or other nuclear processes. Anti-Histone H4 pan Antibody is ideal for research in Chromatin Remodeling, Gene Expression and Epigenetics.

Anti-Histone H4 pan Antibody is suitable for Western Blot, Chromatin Immunoprecipitation, Dot Blots, and ELISA. Expect a single band approximately 11.3 kDa in size corresponding to H4 by western blot in the appropriate tissue or cell lysate. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty