Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAG Mouse Monoclonal Antibody (BAC) [Clone: [7F4]]

Supplier: Rockland Immunochemicals
200-306-BR6
75928-484EA 539 USD InStock
75928-484
Anti-GAG Mouse Monoclonal Antibody (BAC) [Clone: [7F4]]
Antibodies
The human immunodeficiency virus type 1 (HIV-1) particle consists of an envelope, a core and the region between the two termed matrix (1). The HIV-1 Gag protein is a late structural protein that contains four proteins: matrix (p17), capsid (p24), nucleocapsid (p7) and the p6 protein (2). The p24 constitutes the major core component of the virus and shows high degree of sequence conservation among HIV isolates. The Gag p24 has been used as an integral part of multicomponent HIV-1 vaccines (3).

Anti-HIV-1 p24 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: GAG
Clonality: Monoclonal
Clone: [7F4]
Conjugation: BAC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty