Would you like to log in again?
Horse Blood In Sodium Citrate, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
R207-0050
75928-520EA 241 USD InStock
75928-520
Horse Blood In Sodium Citrate, Rockland Immunochemicals
Blood
Horse Blood In Sodium Citrate
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Citrated horse blood is aseptically collected and prepared to order from our colony of animals. Citrated horse blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Citrated Horse RBCs are also useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Horse red blood cells are perishable and are collected and processed upon receipt of your order.
Ideal for production of blood agar plates, washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty