Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HRP Western Blot Anti-Mouse IgG Polyclonal Antibody

Supplier: Rockland Immunochemicals
KCB002
75928-536EA 112 USD InStock
75928-536
Anti-HRP Western Blot Anti-Mouse IgG Polyclonal Antibody
Antibodies
Anti-Mouse IgG Antibody Peroxidase Conjugated for Western Blot

  • prepared from normal serum by a multi-stage process which includes delipidation, salt fractionation and ion exchange chromatography
  • Use with KCA002 - Chemiluminescent Western Blot Kit for use with Mouse Primary Antibody FemtoMax™

HRP is used in applications primarily for its ability to amplify a weak signal and increase detectability of a target molecule. HRP-conjugated secondary antibodies are utilized in conjunction with specific chemiluminescent substrates to generate the light signal and HRP conjugates have a very high turnover rate, yielding good sensitivity with short reaction times. Chemiluminescence is the best all-around method for western blot detection as these systems eliminate the hazards associated with radioactive materials and toxic chromogenic substrates. These methods are also unmatched in the speed and sensitivity compared to traditional alternatives, utilizing film to record and store data permanently. Blots detected with chemiluminescent methods are easily stripped for subsequent reprobing with additional antibodies. This item is a component of kit (KCA002). The Kit contains all the reagents necessary to perform chemiluminescent Western Blots to detect mouse primary antibodies.

HRP Western Blot Anti-Mouse IgG Antibody is specifically designed for immunoblotting used with Chemiluminescent Western Blot Kit (p/n KCA002) with Mouse Primary Antibody.

Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty