Would you like to log in again?
Hamster Sciatic Nerve, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
HM-T175
75928-688EA 82 USD InStock
75928-688
Hamster Sciatic Nerve, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Hamster Sciatic Nerve
  • Harvested Animal Tissue
Fresh-Frozen collection of tissue
healthy non-immunized animals
useful for staining assays, controls, lysates, sequencing
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty