Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1RL2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BV4
75928-872EA 518 USD InStock
75928-872
Anti-IL1RL2 Rabbit Polyclonal Antibody
Antibodies
IL-1RL2 is a member of the interleukin 1 receptor family, but it is incapable of binding to interleukin 1 alpha and interleukin 1 beta with high affinity (1). Together with IL-1RAcP, it can bind members of the IL-36 cytokine family, leading to activation of the NF-kappaB pathway (2). IL-1RL2 can also bind to IL-1F10, resulting in a decreased product of Th17 cytokines in response to immunological or LPS challenge, suggesting that one potential role of IL-1RL2 may be to modulate the immune and inflammation response (3).

Anti-IL-1RL2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 65 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IL1RL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty