Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL23A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-G99
75928-888EA 518 USD InStock
75928-888
Anti-IL23A Rabbit Polyclonal Antibody
Antibodies
IL-23 antibody detects human IL-23. Like interleukin-27 (IL-27), IL-23 is a recently discovered member of the IL-6/IL-12 family of proinflammatory and immunoregulatory cytokines. It exists as a heterodimer composed of the IL-12p40 subunit and a novel p19 subunit. IL-23 is secreted by activated dendritic cells, macrophages, and monocytes. Its biological activities include enhancing the proliferation of memory T cells and the production of IFN-gamma, IL-12, and TNF-alpha from activated T cells, and can stimulate macrophages to produce TNF-alpha and nitric oxide. It has also been shown to possess potent anti-tumor and anti-metastatic activity in mouse models of cancer, suggesting a potential role for IL-23 in therapeutic treatment of cancer. Anti-IL-23 antibodies are ideal for investigators involved in cytokine and growth factors and cancer research.

Anti-IL-23 Antibody is suitable for use in E,WB, and ICC. Expect a band approximately ~20.7 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: IL23A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty