Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL37 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BX7
75928-924EA 518 USD InStock
75928-924
Anti-IL37 Rabbit Polyclonal Antibody
Antibodies
Interleukin 37 (IL-37) is a member of the interleukin 1 cytokine family (reviewed in 1). It can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). IL-37 also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18 (1). IL-37 can also interact with Smad3 and expression of IL-37 in macrophages or epithelial cells resulted in near complete suppression of pro-inflammatory cytokines, suggesting that IL-37 may act as a natural suppressor of the innate inflammatory and immune responses (2,3).

Anti-IL-37 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 24 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IL37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty