Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C3orf54 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BY2
75928-934EA 518 USD InStock
75928-934
Anti-C3orf54 Rabbit Polyclonal Antibody
Antibodies
INCA1 was isolated through a yeast triple hybrid approach to identify interaction partners of the cyclin A1-CDK2 heterodimer. INCA1 is evolutionarily conserved and has no homology to any known proteins. Like Cyclin A1, INCA1 is highly expressed in testis with highest levels observed at various points of testis maturation. It has been suggested that the complex of INCA1 and the cyclinA1-CDK2 dimer plays a role in several signaling pathways important for cell cycle control and meiosis. Recent experiments have shown that INCA1 also interacts with the testis protein RSB-66 and this interaction takes place in the cytoplasm.

Anti-INCA1 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 31 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: C3orf54
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty