Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SUC1 Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-189S
75929-186EA 148 USD InStock
75929-186
Anti-SUC1 Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Invertase antibody recognizes invertase which hydrolyzes the alpha-1,4-glycosidic bond between alpha-D-glucose and beta-D-fructose of sucrose. Bees use invertase to convert nectar to honey. In the confectionary industry, yeast-derived invertase is used to hydrolyze sucrose to glucose and fructose as fructose is more sweeter and less prone to crystallization.

Anti-Invertase antibody is ideal for investigators interested in Metabolism and Cell Biology.

Anti-Invertase antibody has been assayed against 1.0 ug of Invertase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: SUC1
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty