Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDM5C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-W42
75929-276EA 487 USD InStock
75929-276
Anti-KDM5C Rabbit Polyclonal Antibody
Antibodies
Jarid1c is a histone demethylase which specifically demethylates lysine 4 of histone H3, thereby playing a internal role in the regulation of transcription and chromatin remodelling. Jarid1c demethylates the trimethylated and dimethylated lysine 4, but not the monomethylated form. Further, Jarid1c participates in transcriptional repression of neuronal genes by recruiting histone deacetylases and REST at neuron-restrictive silencer elements. Mutations in the Jarid1c gene have been associated with X-linked mental retardation. Anti-Jarid1c Antibody is ideal for research in Epigenetics, Gene Expression and Neuroscience.

Anti-Jarid1c Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 175 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: KDM5C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty