Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDM3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CB3
75929-314EA 518 USD InStock
75929-314
Anti-KDM3A Rabbit Polyclonal Antibody
Antibodies
The jumonji domain containing 1A protein (JMJD1A) was originally discovered as a testes specific gene, but has been found to be expressed in numerous tissues. JMJD1A is a histone demethylase and specifically demethylates mono- and dimethyl-H3K9. It has also been found to be a novel interaction partner with ER71, a transcription factor expressed in the testes of adult mice and during embryogenesis. JMJD1A is also a downstream gene of STAT3, a protein that has been found to be important in the maintenance of mouse embryonic stem (mES) cells, and decreased JMJD1A expression correlated with the differentiation of cultured mES cells following the removal of the cytokine LIF. These findings suggest that JMJD1A might be a critical signaling molecule underlying the maintenance of pluripotency in embryonic stem cells. At least two isoforms of JMJD1A are known to exist.

Anti-JMJD1A Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 148 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: KDM3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty