Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDM4B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V90
75929-326EA 518 USD InStock
75929-326
Anti-KDM4B Rabbit Polyclonal Antibody
Antibodies
JMJD2b, belongs to the JMJD2 family of histone demethylases which play an important role in the establishment of the histone code. JMJD2b specifically demethylates the trimethylated K9 of histone H3. It is not able to demethylate K4, K27 and K36 of histone H3, nor K20 of histone H4. Anti-JMJD2b Antibody is ideal for research in Epigenetics and Chromatin Remodeling.

Anti-JMJD2b Antibody is suitable for ELISA, Immunofluorescence and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 120 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: KDM4B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty