Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KEAP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CE1
75929-434EA 518 USD InStock
75929-434
Anti-KEAP1 Rabbit Polyclonal Antibody
Antibodies
KEAP1 (kelch-like ECH-associated protein 1) is a stress sensing adaptor for the Cullin3 (Cul3)-dependent E3 ubiquitin ligase complex that negatively regulates NRF2 (NF-E2-related factor 2) and plays a role in the oxidative stress response. It targets NFE2L2/NRF2 for ubiquitination and degradation by the proteasome. KEAP1 contains an amino terminal BTB/POZ domain and a carboxyl terminal KELCH domain which are required for interaction with NRF2, and in binding Cul3-E3 ubiquitin ligase. Altered expression of NRF2 is associated with chronic obstructive pulmonary disease (COPD). KEAP1 also targets the down regulation of NF-kB activity by targeting IKKb degradation. Mutation of the KEAP1 gene is found in lung cancer.

Anti-KEAP1 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 70 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: KEAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty