Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLK4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CF3
75929-450EA 518 USD InStock
75929-450
Anti-KLK4 Rabbit Polyclonal Antibody
Antibodies
Kallikreins (KLKs) belong to the serine protease family of proteolytic enzymes having diverse physiological functions (1). Kallikrein 4 (KLK4), also known as prostase or EMSP1 (enamel matrix serine protease 1), contains one peptidase S1 domain and the expression in prostate is regulated by hormone (1,2). Many kallikreins are implicated in carcinogenesis and have potential as novel cancer and other disease biomarkers (3). KLK4 may have additional roles such as functioning as one of the two major enamel proteases identified that process enamel matrix proteins. Defects in KLK4 are the cause of amelogenesis imperfecta hypomaturation type 2A1 (AI2A1) (4).

Anti-KLK4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 27 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: KLK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty