Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KPNA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CF9
75929-462EA 518 USD InStock
75929-462
Anti-KPNA1 Rabbit Polyclonal Antibody
Antibodies
Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA1, also known as importin alpha 5, is the alpha subunit of karyopherin, which forms a complex with importin subunit beta-1 and functions in nuclear protein import as an adapter protein for nuclear receptor KPNB1. It is ubiquitously expressed and polyubiquitinated in the presence of RAG1. KPNA1 interacts with various virus nucleoproteins, including those of Ebola and influenza.

Anti-KPNA1 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 60 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: KPNA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty