Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-N Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CH0
75929-486EA 518 USD InStock
75929-486
Anti-N Rabbit Polyclonal Antibody
Antibodies
La Crosse virus (LCV) is a single-stranded RNA virus that can cause encephalitis in humans and is transmitted predominantly by mosquitoes. It is often mistaken for herpes simplex encephalitis (1). During human infections, LCV initially presents as a nonspecific summertime illness with fever, headache, nausea, vomiting and lethargy. Death occurs in less than 1% of clinical cases.

Anti-La Crosse Virus Nucleocapsid Antibody has been tested for use in ELISA. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 27 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: N
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty