Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAMTOR1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CH3
75929-520EA 518 USD InStock
75929-520
Anti-LAMTOR1 Rabbit Polyclonal Antibody
Antibodies
LAMTOR1 is a 161 amino acid membrane protein belonging to the LAMTOR family. It has been identified to interact with and recruit the four Rag GTPases (Rag A-D) to the surface of lysosomes (1). The mTORC1 kinase complex is a critical component in the regulation of cell growth (2). As part of the ragulator complex, LAMTOR1 recruits the Rag GTPases and the mTORC1 complex to lysosomes, a key step in activation of the TOR signaling cascade by amino acids (3). LAMTOR1 may be involved in cholesterol homeostasis regulating LDL uptake and cholesterol release from late endosomes / lysosomes (4).

Anti-LAMTOR1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 18 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LAMTOR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty