Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAR/PTPRF Mouse Monoclonal Antibody (HRP) [Clone: S165-38]

Supplier: Rockland Immunochemicals
200-303-W57
75929-550EA 539 USD InStock
75929-550
Anti-LAR/PTPRF Mouse Monoclonal Antibody (HRP) [Clone: S165-38]
Antibodies
PTPRF or leukocyte common antigen-related protein (LAR) is a widely expressed protein tyrosine phosphatase with an extracellular receptor region that resembles a cell adhesion molecule. PTPRF removes phosphate group from β-catenin, an event that may subsequently facilitate cell-cell adhesion and ensure the stability of the cadherin complex. This phosphatase has also been implicated in various cellular processes such as neurite growth, nerve regeneration, actin remodeling and regulation of insulin function (1,2,3,4). Anti-PTPRF (C-terminal) antibody is specific for the extracellular and cytoplasmic subunits of human PTPRF (approx. 210, 150 and 85 kDa). Detection of the PTPRF bands by immunoblotting is specifically inhibited by the immunizing peptide. Anti-PTPRF is ideal for researchers interested in Cell adhesion Cadherin-mediated cell adhesion pathways, PAK pathways, insulin resistance and ureterocele.

Anti-LAR/PTPRF HRP Conjugated Antibody is suitable for use in Western blots and Immunocytochemistry. Expect a band approximately ~85 kDa on specific lysates or tissues. Full protein is 210 kDa; degrades due to proteolysis into p-subunit containing transmembrane and intracellular domains. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: LAR/PTPRF
Clonality: Monoclonal
Clone: S165-38
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty