Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LEMD3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CJ3
75929-574EA 518 USD InStock
75929-574
Anti-LEMD3 Rabbit Polyclonal Antibody
Antibodies
LEMD3 (LEM domain-containing 3) is a lem-domain containing protein, also known as MAN1, whose loss of function results in disorders characterized by osteopoikilosis, Buschke-Ollendorff syndrome, and melorheostosis. LEMD3 is an inner nuclear membrane protein that interacts with BMP and activin-TGF-beta receptor-activated Smads and antagonizes TGF-beta signaling in human cells.

Anti-LEMD3 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 100 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LEMD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty