Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LGI3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CJ8
75929-618EA 518 USD InStock
75929-618
Anti-LGI3 Rabbit Polyclonal Antibody
Antibodies
The leucine-rich, glioma inactivated gene 3 (LGI3) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI3 have not been found to be associated with ADTLE. LGI3 expression is induced in rat astrocyte cultures by the amyloid beta (Abeta) peptide and accumulated on neuronal plasma membranes of aged monkey brains and co-localized with Abeta.

Anti-LGI3 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 62 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LGI3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty