Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CK5
75929-632EA 518 USD InStock
75929-632
Anti-LIF Rabbit Polyclonal Antibody
Antibodies
LIF is a pleiotropic cytokine with roles in several different systems. It is involved in the induction of hematopoietic differentiation in normal and myeloid leukemia cells, induction of neuronal cell differentiation, regulator of mesenchymal to epithelial conversion during kidney development, and may also have a role in immune tolerance at the maternal-fetal interface. LIF was initially recognized by its ability to induce terminal differentiation of myeloid leukemic cells. It is a member of the IL-6 cytokine superfamily and can be highly glycosylated. LIF signaling is transduced through the LIF-R/gp130 receptor complex, leading to the phosphorylation and activation of the JAK/STAT pathway. Recent evidence shows that LIF inhibits cardiomyogenesis in embryonic stem cells via STAT3 activation.

Anti-LIF Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 22 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LIF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty