Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIMA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CK6
75929-634EA 518 USD InStock
75929-634
Anti-LIMA1 Rabbit Polyclonal Antibody
Antibodies
The Lim domain and actin-binding protein 1 (LIMA1) is cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles and is downregulated in some cancer cell lines (1,2). LIMA1 is also a key molecule linking the cadherin-catenin complex to the actin cytoskeleton (3). Recent studies have shown that EGF activates ERK1/2-dependent phosphorylation, ubiquitination and degradation of LIMA1, leading to increased invasiveness and metastasis of metastatic prostate cancer models (4).

Anti-LIMA1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 85 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LIMA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty