Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIN54 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CL1
75929-644EA 518 USD InStock
75929-644
Anti-LIN54 Rabbit Polyclonal Antibody
Antibodies
LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes. This complex functions as a transcriptional repressor by binding to promoters of E2F target genes that regulate the G1/S transition (1). LIN54 is required for cell cycle progression and can bind to the cdc2 promoter in a sequence-specific manner (2).

Anti-LIN54 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 79 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LIN54
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty