Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LIPE Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H01
75929-646EA 518 USD InStock
75929-646
Anti-LIPE Rabbit Polyclonal Antibody
Antibodies
Lipe antibody detects human Lipe. Although initially described as an adipocyte-specific triacylglycerol lipase, lipe (also known as hormone-specific lipase or HSL) is expressed in multiple tissues and cell lines. It plays multiple roles in lipid metabolism, including hormone-stimulated lipolysis in adipose tissue and the hydrolysis of cholesterol esters. Lipe is expressed as a long and a short form, generated by use of alternative translational start codons. The long form is expressed in steroidogenic tissues such as testis, where it converts cholesterol esters to free cholesterol for steroid hormone production. The short form is expressed in adipose tissue, among others, where it hydrolyzes stored triglycerides to free fatty acids. Recently, it was observed that the lack of lipe in genetically obese leptin-null mice inhibited obesity and adipogenesis, suggesting that lipe plays a major role in adipocyte proliferation. Anti-Lipe antibodies are ideal for investigators involved in Apolipoprotein reseach.

Anti-Lipe Antibody is suitable for use in E,WB, and IHC. Expect a band approximately ~116.5 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: LIPE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty