Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRRFIP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CM7
75929-690EA 518 USD InStock
75929-690
Anti-LRRFIP1 Rabbit Polyclonal Antibody
Antibodies
LRRFIP1, also known as GC-binding factor 2 (GCF2), is a 738 amino acid transcriptional repressor that mainly plays a cytoskeletal role. It is primarily localized in the cytoplasm and preferentially binds to GC-rich dsDNA, but will also bind directly to dsRNA as well. The RNA binding domain encompasses a lysine-rich motif. LRRFIP1 interacts with the mammalian Flightless I (Fli-I) and is a key component in the cytoskeletal regulation of platelet function. LRRFIP1 and the related protein LRRFIP2 may modulate canonical WNT signaling and mediate the IRF3-induced production of type I interferon via a beta-catenin-dependent pathway.

Anti-LRRFIP1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 90 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LRRFIP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty