Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRRK2 Mouse Monoclonal Antibody (FITC) [Clone: S231B-34]

Supplier: Rockland Immunochemicals
200-302-W59
75929-726EA 539 USD InStock
75929-726
Anti-LRRK2 Mouse Monoclonal Antibody (FITC) [Clone: S231B-34]
Antibodies
LRRK2 is a large protein with multiple domains including several ankyrin, leucine-rich, and WD40 repeats, a Ras-like small GTPase family domain named Roc, and a kinase domain that is closely related to the RIP kinase domain. LRRK2 gene is expressed in brain as well as in other tissues such as lung, liver and heart. LRRK2 might be central to the pathogenesis of several major neurodegenerative diseases associated with parkinsonism. Several dominantly inherited missense mutations in the gene encoding LRRK2 have been identified in several families that exhibit a broad spectrum of neuropathological features. Anti-LRRK2 is ideal for researchers interested in Neuroscience, specifically parkinsonism, as well as pathways related to the Electron Transport Chain and Wnt Signaling.

Anti-LRRK2/Dardarin FITC Conjugated Antibody is suitable for Western Blots, Immunocytochemistry, and Immunohistochemistry. Expect a band approximately >200 kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: LRRK2
Clonality: Monoclonal
Clone: S231B-34
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty