Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LZTS2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CP5
75929-770EA 518 USD InStock
75929-770
Anti-LZTS2 Rabbit Polyclonal Antibody
Antibodies
The leucine zipper putative tumor suppressor 2 (LZTS2, also known as LAPSER1), a member of the LZTS family, is a negative regulator of the Wnt signaling pathway. It represses beta-catenin-mediated transcriptional activation by promoting the nuclear exclusion of beta-catenin. LZTS2 is involved in cytokinesis and regulates katanin-mediated microtubule severing and release from the centrosome. LZTS2 may negatively regulate axonal outgrowth by preventing the formation of microtubule bundles that are necessary for transport within the elongating axon. LZTS2 is highly expressed in testis and prostate, but it is deleted in multiple cancers, including prostate tumors, suggesting that LZTS2 acts as a potential tumor suppressor.

Anti-LZTS2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 73 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LZTS2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty